Benzyloxyacetic acid ≥97.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

B2876-1G B2876-5G
TCIAB2876-1GEA 17.6 EUR
TCIAB2876-1G TCIAB2876-5G
Benzyloxyacetic acid ≥97.0% (by GC, titration analysis)
Benzyloxyacetic acid
Kaava: C₉H₁₀O₃
Kiehumispiste: 155°C
MDL numero: MFCD00274211
CAS-numero: 30379-55-6

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR