2-Bromo-3-formylpyridine ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

B2625-1G B2625-5G
TCIAB2625-1GEA 38.5 EUR
TCIAB2625-1G TCIAB2625-5G
2-Bromo-3-formylpyridine ≥98.0% (by GC)
2-Bromo-3-formylpyridine
Kaava: C₆H₄BrNO
Kiehumispiste: 100°C
MDL numero: MFCD04966945
CAS-numero: 128071-75-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=895a0d341695e9577ace77ba4cc28b54a1719249&vid=895a0d341695e9577ace77ba4cc28b54a1719249&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR