Litiumbis(trifluorimetyylisulfonyyli)imidi ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B2542-25G B2542-250G
TCIAB2542-25GEA 80.3 EUR
TCIAB2542-25G TCIAB2542-250G
Litiumbis(trifluorimetyylisulfonyyli)imidi ≥98.0% (by titrimetric analysis)
Litiumbis(trifluorimetyylisulfonyyli)imidi
Kaava: C₂F₆LiNO₄S₂
MDL numero: MFCD00210017
CAS-numero: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2338b0093fc16d63b192fc41e50b037cf83f1dc1&vid=2338b0093fc16d63b192fc41e50b037cf83f1dc1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 231.0 to 237.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR