1-Boc-4-(hydroxymethyl)piperidine ≥98.0% (by GC)

Toimittaja: TCI

B2364-1G B2364-25G B2364-5G
TCIAB2364-1GEA 29.7 EUR
TCIAB2364-1G TCIAB2364-25G TCIAB2364-5G
1-Boc-4-(hydroxymethyl)piperidine ≥98.0% (by GC)
1-Boc-4-(hydroxymethyl)piperidine
Kaava: C₁₁H₂₁NO₃
MDL numero: MFCD02094488
CAS-numero: 123855-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04916a4bcfc763b92db30a1adcdbc983e80c7621&vid=04916a4bcfc763b92db30a1adcdbc983e80c7621&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR