tert-Butyl (2-bromoethyl)carbamate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

B2289-1G B2289-25G B2289-5G
TCIAB2289-1GEA 46.2 EUR
TCIAB2289-1G TCIAB2289-25G TCIAB2289-5G
tert-Butyl (2-bromoethyl)carbamate ≥98.0% (by GC)
tert-Butyl (2-bromoethyl)carbamate
Kaava: C₇H₁₄BrNO₂
Storage Temperature: Pakastin
MDL numero: MFCD02683428
CAS-numero: 39684-80-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR