Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI

B2253-25G B2253-5G
TCIAB2253-25GEA 80.3 EUR
TCIAB2253-25G TCIAB2253-5G
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Kaava: C₁₈H₂₆N₂O₆
MDL numero: MFCD00038259
CAS-numero: 2480-93-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 92.0 to 95.0 deg-C
Specific rotation [a]20/D -5.0 to -1.0 deg (C=1, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR