[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B2226-1G B2226-5G
TCIAB2226-1GEA 57.2 EUR
TCIAB2226-1G TCIAB2226-5G
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥97.0% (by titrimetric analysis)
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride
Kaava: C₃₄H₂₈Cl₂FeNiP₂
MDL numero: MFCD00270284
CAS-numero: 67292-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5298d613ceac809bdc5bacd6c6f60bc77171d2bd&vid=5298d613ceac809bdc5bacd6c6f60bc77171d2bd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Green to Dark green powder to crystal
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR