Boc-D-Arg-OH·HCl·H2O ≥98.0% (by total nitrogen basis)

Toimittaja: TCI

B2204-1G B2204-5G
TCIAB2204-1GEA 24.2 EUR
TCIAB2204-1G TCIAB2204-5G
Boc-D-Arg-OH·HCl·H2O ≥98.0% (by total nitrogen basis)
Boc-D-Arg-OH·HCl·H2O
Kaava: C₁₁H₂₂N₄O₄·HCl·H₂O
MDL numero: MFCD00065554
CAS-numero: 204070-00-8

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0% (calcd.on anh.substance)
Specific rotation [a]20/D +6.0 to +10.0 deg (C=2,water)
Solubility in Water very faint turbidity
Water 4.5 to 6.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR