2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

B2144-25G B2144-5G
TCIAB2144-25GEA 208 EUR
TCIAB2144-25G TCIAB2144-5G
2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)
2,2,2-Trifluoroethyl butyrate
Kaava: C₆H₉F₃O₂
Leimahduspiste: 24°C
MDL numero: MFCD00077609
CAS-numero: 371-27-7
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc449a0680aa837f0cc8f94287f57f9e2d07dcb5&vid=dc449a0680aa837f0cc8f94287f57f9e2d07dcb5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR