3-Bromo-4-fluorobenzonitrile ≥99.0% (by GC)

Toimittaja: TCI
Vaara

B1965-25G B1965-5G
TCIAB1965-25GEA 90.2 EUR
TCIAB1965-25G TCIAB1965-5G
3-Bromo-4-fluorobenzonitrile ≥99.0% (by GC)
3-Bromo-4-fluorobenzonitrile
Kaava: C₇H₃BrFN
Kiehumispiste: 134 - 136 °C (33 -rr)
Sulamispiste: 53…58 °C
MDL numero: MFCD00055432
CAS-numero: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39d5705402e001d94f57934addaa34459576ac85&vid=a31249149b6ba10b38a63e1ee883396421eeabf5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 55.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR