2,2'-Bithiophene-5-carboxaldehyde ≥98.0% (by GC)

Toimittaja: TCI

B1838-1G B1838-25G B1838-5G
TCIAB1838-1GEA 49.5 EUR
TCIAB1838-1G TCIAB1838-25G TCIAB1838-5G
2,2'-Bithiophene-5-carboxaldehyde ≥98.0% (by GC)
2,2'-Bithiophene-5-carboxaldehyde
Kaava: C₉H₆OS₂
Kiehumispiste: 210°C
MDL numero: MFCD00052294
CAS-numero: 3779-27-9

Order Now

Testitulosten erittely

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR