Bis(triphenylphosphine)nickel(II) chloride ≥96.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B1571-10G B1571-100G
TCIAB1571-10GEA 34.1 EUR
TCIAB1571-10G TCIAB1571-100G
Bis(triphenylphosphine)nickel(II) chloride ≥96.0% (by titrimetric analysis)
Bis(triphenylphosphine)nickel(II) chloride
Kaava: C₃₆H₃₂Cl₂NiP₂
MDL numero: MFCD00009592
CAS-numero: 14264-16-5
EINECS: 238-154-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5798ae79f9085fa172f51f825c8d503ce47e02c4&vid=5798ae79f9085fa172f51f825c8d503ce47e02c4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Chelometric Titration) min. 96.0 %
Purity (Iodometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR