Ethyl-tert-butylacetate ≥99.0% (by GC)

Toimittaja: TCI
Varoitus

B1510-100ML B1510-25ML
TCIAB1510-100MLEA 273 EUR
TCIAB1510-100ML TCIAB1510-25ML
Ethyl-tert-butylacetate ≥99.0% (by GC)
Ethyl-tert-butylacetate
Kaava: C₈H₁₆O₂
Kiehumispiste: 145°C
Leimahduspiste: 34°C
MDL numero: MFCD00026911
CAS-numero: 5340-78-3
EINECS: 226-280-6
UN: 3272
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR