N-tert-Butyl-2-methoxyethylamine ≥98.0% (by GC)

Toimittaja: TCI
Vaara

B1430-1G B1430-5G
TCIAB1430-1GEA 45.1 EUR
TCIAB1430-1G TCIAB1430-5G
N-tert-Butyl-2-methoxyethylamine ≥98.0% (by GC)
N-tert-Butyl-2-methoxyethylamine
Kaava: C₇H₁₇NO
Kiehumispiste: 137°C
MDL numero: MFCD00145008
UN: 2733
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba210b6d3277aa7aea37ef8cea76bd5d565fd64a&vid=2a50d141ccbd7d34762ce1160c8a54f415c03532&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR