1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B1313-25G B1313-5G
TCIAB1313-25GEA 229 EUR
TCIAB1313-25G TCIAB1313-5G
1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0% (by titrimetric analysis)
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
Kaava: C₂₇H₂₆Cl₂NiP₂
MDL numero: MFCD00015318
CAS-numero: 15629-92-2

Order Now

Testitulosten erittely

Appearance Orange to Brown to Dark red powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR