Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis) for organic electronics

Toimittaja: TCI

B1221-25G B1221-5G
TCIAB1221-25GEA 501 EUR
TCIAB1221-25G TCIAB1221-5G
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis) for organic electronics
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex
Kaava: C₃₈H₇₂N₂S₁₀Zn
MDL numero: MFCD00060267
CAS-numero: 68449-38-7

Order Now

Testitulosten erittely

Appearance Red to Dark red to Dark purple powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Chelometric Titration) min. 98.0 %
Melting point 170.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR