tert-Butylacetyl chloride ≥99.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B1196-25ML B1196-250ML
TCIAB1196-25MLEA 45.1 EUR
TCIAB1196-25ML TCIAB1196-250ML
tert-Butylacetyl chloride ≥99.0% (by titrimetric analysis)
tert-Butylacetyl chloride
Kaava: C₆H₁₁ClO
Kiehumispiste: 132°C
Leimahduspiste: 21°C
MDL numero: MFCD00000737
CAS-numero: 7065-46-5
EINECS: 230-356-4
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee4484562913d07b3f462827d3286f6f12e23ca3&vid=ee4484562913d07b3f462827d3286f6f12e23ca3&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Red to Green clear liquid
Purity (Argentmetric Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR