1,4-Bis(2-hydroxyethoxy)-2-butyne >50.0% (by GC)

Toimittaja: TCI

B0979-25G
TCIAB0979-25GEA 53.9 EUR
TCIAB0979-25G
1,4-Bis(2-hydroxyethoxy)-2-butyne >50.0% (by GC)
1,4-Bis(2-hydroxyethoxy)-2-butyne
Kaava: C₈H₁₄O₄
MDL numero: MFCD00002883
CAS-numero: 1606-85-5
EINECS: 216-526-0

Order Now

Testitulosten erittely

Appearance Colorless to Brown clear liquid
Purity (GC) min. 50.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR