N-tert-Butyl-5-methylisoxazolium Perchlorate ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI
Varoitus

B0832-1G B0832-5G
TCIAB0832-1GEA 83.6 EUR
TCIAB0832-1G TCIAB0832-5G
N-tert-Butyl-5-methylisoxazolium Perchlorate ≥98.0% (by HPLC, total nitrogen)
N-tert-Butyl-5-methylisoxazolium Perchlorate
Kaava: C₈H₁₄ClNO₅
MDL numero: MFCD00011960
CAS-numero: 10513-45-8
EINECS: 234-048-0
UN: 1479
ADR: 5.1,III

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 119.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR