4,4’-Bis(dimetyyliamino)bentsofenoni ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

B0481-25G B0481-500G B0481-100G
TCIAB0481-25GEA 25.3 EUR
TCIAB0481-25G TCIAB0481-500G TCIAB0481-100G
4,4’-Bis(dimetyyliamino)bentsofenoni ≥98.0% (by GC, titration analysis)
4,4’-Bis(dimetyyliamino)bentsofenoni
Kaava: C₁₇H₂₀N₂O
Kiehumispiste: 428 °C (1013 hPa)
Sulamispiste: 172 °C
Leimahduspiste: 220 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008312
CAS-numero: 90-94-8
EINECS: 202-027-5
ADR: 6.1,III
Merck Index: 14,06177
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2bd084b7efbb27eb705bad5e93639018bba03d4&vid=c2bd084b7efbb27eb705bad5e93639018bba03d4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR