Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

B0457-25G B0457-500G
TCIAB0457-25GEA 22 EUR
TCIAB0457-25G TCIAB0457-500G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Girard's reagent T (carbazoylmethyltrimethylammonium chloride)
Kaava: C₅H₁₄ClN₃O
MDL numero: MFCD00012009
CAS-numero: 123-46-6
EINECS: 204-629-3
Merck Index: 14,04429
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb6a70d7bc4ba8944f6e07365e918fb4307b57e8&vid=bb6a70d7bc4ba8944f6e07365e918fb4307b57e8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR