Dibentsyylisulfidi ≥98.0% (by GC)

Toimittaja: TCI

B0440-25G
TCIAB0440-25GEA 29.7 EUR
TCIAB0440-25G
Dibentsyylisulfidi ≥98.0% (by GC)
Dibentsyylisulfidi
Kaava: C₁₄H₁₄S
Kiehumispiste: 131°C
Sulamispiste: 47…50 °C
Tiheys: 1,0712 g/cm³ (50 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00004784
CAS-numero: 538-74-9
EINECS: 208-703-6
ADR: 9,
Merck Index: 14,01145
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bd8fc21110c9bf9a6e10a87a97f9d3a4d0049c6&vid=1bd8fc21110c9bf9a6e10a87a97f9d3a4d0049c6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 50.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR