N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)

Toimittaja: TCI

B0309-1G B0309-100MG
TCIAB0309-1GEA 191 EUR
TCIAB0309-1G TCIAB0309-100MG
N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)
N-Benzoyl-DL-phenylalanine-2-naphthyl ester
Kaava: C₂₆H₂₁NO₃
MDL numero: MFCD00021607
CAS-numero: 15873-25-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 156.0 to 159.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR