(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

A3036-25G
TCIAA3036-25GEA 135 EUR
TCIAA3036-25G
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol ≥98.0% (by HPLC, titration analysis)
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol
Kaava: C₉H₁₂N₂O₄
MDL numero: MFCD00007359
CAS-numero: 2964-48-9
EINECS: 221-001-4

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +31.0 to +34.0 deg (C=1, 6N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR