5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)

Toimittaja: TCI
Varoitus

A2984-1G A2984-5G
TCIAA2984-1GEA 106 EUR
TCIAA2984-1G TCIAA2984-5G
5-Aminovaleric acid Hydroiodide, low water content ≥97.0% (by total nitrogen and titration analysis)
5-Aminovaleric acid Hydroiodide
Kaava: C₅H₁₁NO₂·HI
CAS-numero: 1705581-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=793c0cc9d7699d4dd09aec578fe81487241756f8&vid=793c0cc9d7699d4dd09aec578fe81487241756f8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Water max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR