tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

A2893-1G A2893-5G
TCIAA2893-1GEA 31.9 EUR
TCIAA2893-1G TCIAA2893-5G
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate ≥98.0% (by titrimetric analysis)
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
Kaava: C₁₁H₂₂N₂O₂
MDL numero: MFCD01317792
CAS-numero: 162167-97-7

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR