3-Aminopropyldimethoxymethylsilane ≥97.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

A2628-25ML A2628-100ML
TCIAA2628-25MLEA 34.1 EUR
TCIAA2628-25ML TCIAA2628-100ML
3-Aminopropyldimethoxymethylsilane ≥97.0% (by GC, titration analysis)
3-Aminopropyldimethoxymethylsilane
Kaava: C₆H₁₇NO₂Si
Kiehumispiste: 84°C
MDL numero: MFCD00025614
CAS-numero: 3663-44-3
EINECS: 222-919-8
UN: 3267
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR