Aztreonam ≥98.0% (by HPLC)

Toimittaja: TCI

A2466-5G A2466-500MG
TCIAA2466-5GEA 226 EUR
TCIAA2466-5G TCIAA2466-500MG
Aztreonam ≥98.0% (by HPLC)
Aztreonam
Kaava: C₁₃H₁₇N₅O₈S₂
Sulamispiste: 227°C
MDL numero: MFCD00072145
CAS-numero: 78110-38-0
EINECS: 278-839-9
Merck Index: 14,00925

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D -26.0 to -32.0 deg (C=0.5、H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR