Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI

A2456-25G A2456-5G
TCIAA2456-25GEA 186 EUR
TCIAA2456-25G TCIAA2456-5G
Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)
Alendronate sodium trihydrate
Kaava: C₄H₁₂NaNO₇P₂·3H₂O
Storage Temperature: Pakastin
MDL numero: MFCD01748233
CAS-numero: 121268-17-5
Merck Index: 14,00229

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Drying loss 16.1 to 17.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR