Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI

Synonyms: trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol HCl, trans-4-((2-Amino-3,5-dibromobenzyl)amino)cyclohexanol hydrochloride

A2360-25G A2360-5G
TCIAA2360-25GEA 310 EUR
TCIAA2360-25G TCIAA2360-5G
Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ambroxol hydrochloride
Kaava: C₁₃H₁₈Br₂N₂O.HCl
Sulamispiste: 235°C
MDL numero: MFCD00078932
CAS-numero: 23828-92-4
EINECS: 245-899-2
Merck Index: 14,00386

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR