Ammoniakki ∼4% ∼2.0 mol/L in isopropanol (IPA)

Toimittaja: TCI
Vaara

A2237-500ML A2237-100ML
TCIAA2237-500MLEA 84.7 EUR
TCIAA2237-500ML TCIAA2237-100ML
Ammoniakki ∼4% ∼2.0 mol/L in isopropanol (IPA)
Ammoniakki
Kaava: NH₃
Storage Temperature: Jääkaappi
MDL numero: MFCD00011418
CAS-numero: 7664-41-7
EINECS: 231-635-3
UN: 1219
ADR: 3,II
Merck Index: 14,00492
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b491975e869699a3c81973c7bfe46ca25190b9c1&vid=20432960c963c0f3a85c568a7837bba053092f15&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless clear liquid
Concentration (Neutralization titration) 4.0 to 4.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR