1-Allyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

A2084-5G A2084-25G
TCIAA2084-5GEA 46.2 EUR
TCIAA2084-5G TCIAA2084-25G
1-Allyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Allyl-3-methylimidazolium chloride
Kaava: C₇H₁₁ClN₂
Storage Temperature: Jääkaappi
MDL numero: MFCD07784450
CAS-numero: 65039-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e95dc4f1c7ab05e5271d6ac310a5f3963fa5501&vid=2e95dc4f1c7ab05e5271d6ac310a5f3963fa5501&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR