Atselaiinihappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

A1318-25G A1318-250G
TCIAA1318-25GEA 25.3 EUR
TCIAA1318-25G TCIAA1318-250G
Atselaiinihappo ≥98.0% (by GC, titration analysis)
Atselaiinihappo
Kaava: C₉H₁₆O₄
Kiehumispiste: 306°C
Sulamispiste: 106 °C
Tiheys: 1,0287 g/cm³ (20 °C)
Leimahduspiste: 215 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004432
CAS-numero: 123-99-9
EINECS: 204-669-1
Merck Index: 14,00905
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=431eb9979a797a32adac0ea5ec6b0cfa3cac149b&vid=431eb9979a797a32adac0ea5ec6b0cfa3cac149b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 106.0 to 109.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR