Allyl-p-tolyl ether ≥95.0% (by GC)

Toimittaja: TCI

A1287-25G A1287-5G
TCIAA1287-25GEA 321 EUR
TCIAA1287-25G TCIAA1287-5G
Allyl-p-tolyl ether ≥95.0% (by GC)
Allyl-p-tolyl ether
Kaava: C₁₀H₁₂O
MDL numero: MFCD00026098
CAS-numero: 23431-48-3

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR