Aminomalononitrile-p-toluenesulphonate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

A1119-25G A1119-5G
TCIAA1119-25GEA 252 EUR
TCIAA1119-25G TCIAA1119-5G
Aminomalononitrile-p-toluenesulphonate ≥98.0% (by titrimetric analysis)
Aminomalononitrile-p-toluenesulphonate
Kaava: C₃H₃N₃
Storage Temperature: Jääkaappi
MDL numero: MFCD00012848
CAS-numero: 5098-14-6
EINECS: 225-817-1
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR