Propyl acetoacetate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

A0817-25ML A0817-500ML
TCIAA0817-25MLEA 82.5 EUR
TCIAA0817-25ML TCIAA0817-500ML
Propyl acetoacetate ≥98.0% (by GC)
Propyl acetoacetate
Kaava: C₇H₁₂O₃
Kiehumispiste: 194°C
MDL numero: MFCD00059405
CAS-numero: 1779-60-8
EINECS: 217-223-6

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR