Sodium-4-aminoazobenzene-4'-sulphonate ≥98.0% (by HPLC)

Toimittaja: TCI

A0748-500G A0748-25G
TCIAA0748-500GEA 198 EUR
TCIAA0748-500G TCIAA0748-25G
Sodium-4-aminoazobenzene-4'-sulphonate ≥98.0% (by HPLC)
Sodium-4-aminoazobenzene-4'-sulphonate
Kaava: C₁₂H₁₁N₃O₃S
MDL numero: MFCD00035564
CAS-numero: 2491-71-6
EINECS: 219-657-1

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR