2-Amino-4,6-dimethylpyridine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

A0731-25G A0731-5G
TCIAA0731-25GEA 162 EUR
TCIAA0731-25G TCIAA0731-5G
2-Amino-4,6-dimethylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-4,6-dimethylpyridine
Kaava: C₇H₁₀N₂
Kiehumispiste: 231°C
MDL numero: MFCD00006322
CAS-numero: 5407-87-4
EINECS: 226-470-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c7f81abbe9a8e23a7231f4dcbb3fd5d3409353&vid=d8c7f81abbe9a8e23a7231f4dcbb3fd5d3409353&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR