α,α,α-Trifluori-p-toluidiini ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

A0662-25ML A0662-100ML
TCIAA0662-25MLEA 66 EUR
TCIAA0662-25ML TCIAA0662-100ML
α,α,α-Trifluori-p-toluidiini ≥98.0% (by GC, titration analysis)
α,α,α-Trifluori-p-toluidiini
Kaava: C₇H₆F₃N
Kiehumispiste: 117°C
Sulamispiste: 3…8 °C
Tiheys: 1,3 g/cm³ (20 °C)
Leimahduspiste: 86°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00064396
CAS-numero: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ede3203f39fee8ee81a6d9c9179dfb47c7c1733&vid=6ede3203f39fee8ee81a6d9c9179dfb47c7c1733&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR