9-Aminofluorene hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

A0622-25G A0622-5G
TCIAA0622-25GEA 399 EUR
TCIAA0622-25G TCIAA0622-5G
9-Aminofluorene hydrochloride ≥98.0% (by HPLC, titration analysis)
9-Aminofluorene hydrochloride
Kaava: C₁₃H₁₁N
MDL numero: MFCD00012536
CAS-numero: 5978-75-6
EINECS: 227-778-6

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR