Acid orange 5

Toimittaja: TCI

A0576-25G A0576-100G
TCIAA0576-25GEA 62.7 EUR
TCIAA0576-25G TCIAA0576-100G
Acid orange 5
Acid orange 5
Kaava: C₁₈H₁₄N₃NaO₃S
MDL numero: MFCD00038130
CAS-numero: 554-73-4
EINECS: 209-071-4
Merck Index: 14,09776

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Lambda max. 446.0 to 450.0 nm (pH3.0)
Absorbance min. 0.270 (5 mg/L, pH 3.0, 446.0 to 450.0 nm)
Transition interval (pH) (purplish red)1.2 to 3.0 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR