D(-)-arabinoosi ≥99.0% (by HPLC)

Toimittaja: TCI

A0513-25G A0513-100G
TCIAA0513-25GEA 40.7 EUR
TCIAA0513-25G TCIAA0513-100G
D(-)-arabinoosi ≥99.0% (by HPLC)
D(-)-arabinoosi
Kaava: C₅H₁₀O₅
Sulamispiste: 160 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00135608
CAS-numero: 10323-20-3
EINECS: 233-708-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50d784a26c928b88ef23dfd533c830e8b08c54d&vid=c50d784a26c928b88ef23dfd533c830e8b08c54d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 156.0 to 165.0 deg-C
Specific rotation [a]20/D -102 to -106 deg (C=10, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR