2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI

A0336-25G A0336-5G
TCIAA0336-25GEA 99 EUR
TCIAA0336-25G TCIAA0336-5G
2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-4-methylthiazole hydrochloride
Kaava: C₄H₆N₂S
MDL numero: MFCD00035214
CAS-numero: 6142-15-0

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 170.0 to 174.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR