1-(2-Aminoethyl)piperazine ≥99.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

A0304-25ML A0304-500ML
TCIAA0304-25MLEA 17.6 EUR
TCIAA0304-25ML TCIAA0304-500ML
1-(2-Aminoethyl)piperazine ≥99.0% (by GC, titration analysis)
1-(2-Aminoethyl)piperazine
Kaava: C₆H₁₅N₃
Kiehumispiste: 222°C
Sulamispiste: -19°C
Leimahduspiste: 93°C
MDL numero: MFCD00005971
CAS-numero: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c589e487c6e92d3816b0d8327700217c4300aa15&vid=c589e487c6e92d3816b0d8327700217c4300aa15&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR