Microplate readers, Infinite® 200 PRO
Toimittaja: Tecan
92000001 92100001 92200001 92300001 92400001 92500001
TCAT92000001EA 0 EUR
TCAT92000001 TCAT92100001 TCAT92200001 TCAT92300001 TCAT92400001 TCAT92500001
Microplate readers, Infinite® 200 PRO
Kuoppalevylukijat
The Infinite 200 PRO plate reader family offers affordable detection solutions empowered by monochromator- or filter based optics.
  • Quad4 Monochromators for free wavelength selection
  • Spectral scanning in both absorbance and fluorescence modes
  • Optimised detectors for each reading mode
  • Filter optics for TR-FRET, HTRF® and FP reading modes
  • z-focussing as integrated background correction
  • Field upgradeable injectors
  • Supports 6 to 384 well plates
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample in parallel
  • A260/280 purity check