96 Well Plate, RCH CEREX PSAX
Toimittaja: Tecan
615-0101RCH
TCAT615-0101RCHEA 0 EUR
TCAT615-0101RCH
96 Well Plate, RCH CEREX PSAX
Kuoppalevyt Kiinteäfaasiuuttolevyt
A 17 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Ideal for acidic target compounds.
Pakkaus: 1 piece per box