96 Well Plate, RCH CEREX MAESTRO
Toimittaja: Tecan
613-0151RCH
TCAT613-0151RCHEA 0 EUR
TCAT613-0151RCH
96 Well Plate, RCH CEREX MAESTRO
Kuoppalevyt Kiinteäfaasiuuttolevyt
Silica-based sorbent with C18 bonding in proprietary coverage.
  • Reversed-phased
  • Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.