MagicPrep™ NGS System
Toimittaja: Tecan
30186621 30186622 30186623 30186624 30186625 30186626 30186627 30186628 30186629 30191459 30191458 30191460 30191465
TCAT30186621EA 0 EUR
TCAT30186621 TCAT30186622 TCAT30186623 TCAT30186624 TCAT30186625 TCAT30186626 TCAT30186627 TCAT30186628 TCAT30186629 TCAT30191459 TCAT30191458 TCAT30191460 TCAT30191465
MagicPrep™ NGS System
Next Generation Sequencing Instruments
MagicPrep™ NGS system is a new category of automated laboratory equipment that provides push button simplicity for NGS library preparation. The Revelo DNA-Seq Enz for MagicPrep™ NGS library preparation kit provides all the reagents for generating DNA-Seq libraries on the MagicPrep™ NGS system.
  • Simple run setup in 10 minutes
  • Reliable and reproducible
  • True walk-away automation
  • Pre-optimised DNA fragmentation for a broad input range simplifies the workflow
  • Provides even coverage and uniformity across the bacterial genome regardless of GC content
  • High quality