Western blots, pre-made, mouse subcellular fraction specific blot
Toimittaja: G Bioscienes
TB47
TB47EA 2090 EUR
TB47
Western blots, pre-made, mouse subcellular fraction specific blot
Elektroforeesireagenssit Western Blotting Reagents
The subcellular fractions from liver tissue are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse livers.
Each blot contains the following mouse liver subcellular fractions: Whole liver, nuclei, mitochondria, Golgi complex, smooth endoplasmic reticulum, and rough endoplasmic reticulum.