Mouse single tissue blot, single species
Toimittaja: G Bioscienes
TB02 TB05 TB08 TB11 TB14 TB17 TB20 TB23 TB26
TB02EA 696 EUR
TB02 TB05 TB08 TB11 TB14 TB17 TB20 TB23 TB26
Mouse single tissue blot, single species
Elektroforeesireagenssit Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue mouse blots